Order Entry
Export
Orders LinkContactUsLinkComponent
7-Hydroxyindole 97%

7-Hydroxyindole 97%

Bottle, 5 g
Catalog # APOSOR10209-5G
Irritant
Supplier:   Apollo Scientific
7-Hydroxyindole 97%
Catalog # APOSOR10209-5G
Supplier:  Apollo Scientific
CAS Number:  2380-84-9
7-Hydroxyindole 97%

Bottle, 5 g

CAS Number:  2380-84-9
Price
348,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:2380-84-9
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152102
  • Molecular Weight:133.15 g/mol
  • Melting Pt:84…88 °C
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications