5-(3-Bromophenyl)cyclohexane-1,3-dione
Bottle, 250 mg
Catalog # APOSOR300642-250MG
Supplier: Apollo Scientific
5-(3-Bromophenyl)cyclohexane-1,3-dione
Catalog # APOSOR300642-250MG
Supplier: Apollo Scientific
CAS Number: 144128-71-2
Jump to:
Product Details & Documents
- CAS Number:144128-71-2
- Formula:C₁₂H₁₁BrO₂
- MDL Number:MFCD03844399
- Molecular Weight:267.13 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



