Quinaldic acid
Bottle, 100 g
Catalog # APOSOR23072-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
- CAS Number:93-10-7
- Formula:C₁₀H₇NO₂
- MDL Number:MFCD00006752
- Molecular Weight:173.17 g/mol
- Melting Pt:156…159 °C
- Storage Temperature:Ambient
- EINECS:202-218-3
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



