Order Entry
Export
Orders LinkContactUsLinkComponent
Bottles, Square, HDPE

Bottles, Square, HDPE

250 ml, OV 45, HDPE, White, 59×59×129,5 mm
Catalog # RIXI2-0201-0250-45
Bottles, Square, HDPE
Catalog # RIXI2-0201-0250-45
Bottles, Square, HDPE

250 ml, OV 45, HDPE, White, 59×59×129,5 mm

Price
71,40 €
Log in for availability
1 * 20 items
Product Details & Documents

Square bottles made up of HDPE plastic, white.

Specifications
  • Thread:
    OV 45
  • Capacity:
    250 ml
  • Colour:
    White
  • Material:
    HDPE
  • Seq:
    02
  • W×D×H:
    59×59×129,5 mm