Order Entry
Export
Orders LinkContactUsLinkComponent
D(-)-Luciferin ≥95%

D(-)-Luciferin ≥95%

Plastic bottle, 1 g
Catalog # AATB12503
Irritant
Supplier:   AAT Bioquest
D(-)-Luciferin ≥95%
Catalog # AATB12503
Supplier:  AAT Bioquest
CAS Number:  2591-17-5
D(-)-Luciferin ≥95%
D(-)-Luciferin ≥95%

Plastic bottle, 1 g

CAS Number:  2591-17-5
Price
872,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Luciferin is the most popular and versatile bioluminescent substrate.

Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g

Specifications