Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cal-630™ AM

Plastic bottle, 1 mg
Catalog # AATB20532
Irritant
Supplier:   AAT Bioquest
Cal-630™ AM
Catalog # AATB20532
Supplier:  AAT Bioquest
CAS Number:  
Cal-630™ AM
Cal-630™ AM

Plastic bottle, 1 mg

CAS Number:  
Price
1 100,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Calcium measurement is critical for numerous biological investigations.

  • Improved sensitivity
  • Improved calcium response
  • Long-wavelength emission
  • Cell permeant

 : For research use only.

  • Molecular Weight:1282.89 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications