Cal-630™ AM
Plastic bottle, 500 µG
Catalog # AATB20531
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
- Cell permeant
Caution: For research use only.
- Molecular Weight:1282.89 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 µG



