XFD488 NHS Ester fluorescent probe
Plastic vial, 1 mg
Catalog # AATB1812
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
XFD488 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 488 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Caution: For research use only.
- Molecular Weight:643.4 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic vial
- Pk:1 mg



