Orders LinkContactUsLinkComponent
MagaDye™ 588 ddTTP
MagaDye™ 588 ddTTP
MagaDye™ 588-ddTTP, 50 Nmo
Catalog # AATB17065
Supplier:   AAT Bioquest
MagaDye™ 588 ddTTP
Catalog # AATB17065
Supplier:  AAT Bioquest
CAS Number:  
MagaDye™ 588 ddTTP
MagaDye™ 588 ddTTP
MagaDye™ 588-ddTTP, 50 Nmo
CAS Number:  
Price
7 440,00 €
Log in for availability
1 * 50 Nmo
Product Details & Documents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.
Specifications
Specifications
  • Pk:
    50 Nmo
  • Description:
    MagaDye™ 588-ddTTP
Specifications