Orders LinkContactUsLinkComponent
PD-10 Buffer Reservoir
PD-10 Buffer Reservoir
PD-10 buffer reservoir
Catalog # AATB60509
Supplier:   AAT Bioquest
PD-10 Buffer Reservoir
Catalog # AATB60509
Supplier:  AAT Bioquest
PD-10 Buffer Reservoir
PD-10 Buffer Reservoir
PD-10 buffer reservoir
Price
275,00 €
Log in for availability
1 * 10 items
Product Details & Documents
PD-10 Buffer Reservoir provides an easy and rapid way for equilibrating PD-10 columns.
  • Efficient and convenient
  • Simple to use
  • Improves equilibration and elution procedures
Caution: For research use only.
Specifications
  • Description:
    PD-10 buffer reservoir
  • Seq:
    02