Orders LinkContactUsLinkComponent
Inoculating loops and needles
Inoculating loops and needles
10 µl, Blue
Catalog # KART2651
Supplier:   KARTELL
Inoculating loops and needles
Catalog # KART2651
Supplier:  KARTELL
Inoculating loops and needles
Inoculating loops and needles
10 µl, Blue
Price
815,00 €
Log in for availability
1 * 8.000 items
Product Details & Documents
High impact PS, sterile.
  • Choice of either 1 µl or 10 µl loop on one end and needle at the other
  • Sterilised by gamma irradiation
Packaging: 20 per bag.
Specifications
  • Capacity:
    10 µl
  • Colour:
    Blue
  • Seq:
    02