Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Storage box with 50 nails 1 * 1 ST
Catalog # PLANG27750B
Supplier:
PLANO W. PLANNET
Expand image
Storage box with 50 nails 1 * 1 ST
Catalog # PLANG27750B
Supplier:
PLANO W. PLANNET