Order Entry
Export
Orders LinkContactUsLinkComponent
 

Liquid pump sampling tool with glass bottle

Clear glass bottle, 500 ml
Catalog # SAMP5021X-500
Supplier:   Sampling Systems
Liquid pump sampling tool with glass bottle
Catalog # SAMP5021X-500
Supplier:  Sampling Systems
Liquid pump sampling tool with glass bottle
Liquid pump sampling tool with glass bottle

Clear glass bottle, 500 ml

Price
10,10 €
Log in for availability
1 * 1 items
Product Details & Documents

This pump sampler set is ideal for contamination-free liquid sampling.

  • Supplied in a convenient carry case
  • Glass bottle container offers excellent chemical resistance
  • Easy to use
  • Suitable for low to medium viscosity liquids

The liquid pump sampler is ideal for sampling liquids from drums and barrels, and is supplied as a set in a handy case. The kit is designed to sample liquids such as water and oil, and is extremely easy to use.

 : Set includes a plastic hand pump, four (4) 500 ml glass bottles, a 13 m LDPE hose, a tube cutter, and carrying case.

Specifications
  • Capacity:
    500 ml
  • Description:
    Clear glass bottle
  • Seq:
    02