ContactUsLinkComponent
DNA (+) Control CISH Probe is designed to be used for the detection of human Alu repetitive sequences in formalin-fixed, paraffin-em bedded tissue or cells by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCO0008.100UL
Supplier:  Abnova
DNA (+) Control CISH Probe is designed to be used for the detection of human Alu repetitive sequences in formalin-fixed, paraffin-em bedded tissue or cells by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCO0008.100UL
Supplier:  Abnova