ContactUsLinkComponent
Glioma 1p/19q Probe Set is designed for the qualitative detection of deletions involving the human chromosomal region 1p36.31 as wel l as deletions involving the human chromosomal region 19q13.32-q13.33 in formalin-fixed, paraffin-embedded specimens 1 * 1 KIT
Catalog # ABNOKA6905.1KIT
Supplier:  Abnova
Glioma 1p/19q Probe Set is designed for the qualitative detection of deletions involving the human chromosomal region 1p36.31 as wel l as deletions involving the human chromosomal region 19q13.32-q13.33 in formalin-fixed, paraffin-embedded specimens 1 * 1 KIT
Catalog # ABNOKA6905.1KIT
Supplier:  Abnova