Orders LinkContactUsLinkComponent
Cryoboxes, Cardboard
Cryoboxes, Cardboard
Origami, Yellow/purple, 52×134×134 mm
Catalog # TENATE22872
Supplier:   TENAK
Cryoboxes, Cardboard
Catalog # TENATE22872
Supplier:  TENAK
Cryoboxes, Cardboard
Cryoboxes, Cardboard
Origami, Yellow/purple, 52×134×134 mm
Price
118,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Colored cryoboxes possible to orient in different ways for classic colour coding, tracking work in progress, identifying the owner of the box, and monitoring sample thawing.
  • PP coated water-repellent surface
  • Internal printed alpha numeric grid
For space-saving solutions, the cryobox is also available in a convenient flat-packed Origami box format.
Specifications
  • Compartments:
    Origami
  • W×D×H ext.:
    52×134×134 mm
  • Colour:
    Yellow/purple
  • Seq:
    07