Code Corporation CR2700 Barcode Scanner Accessories
Cable, 6' length, dark grey, for model CR2700 Inductive charging station (176514)
Catalog # BRDY176508
Supplier: Brady
Jump to:
Product Details & Documents
Accessories available for the Barcode Scanner CR2700.
Cable (BRDY176508) - Use this 6-foot straight cable for making USB to Micro USB connections. Reader termination: Micro USB. Host termination: USB Type A.
B27 lithium-ion battery (BRDY176699) - powers the CR2700 reader with a unique set of features that keep workflow interruptions to a minimum. Power gauge to track battery level. Quick-release latch allows for toolless swapping. The B27 battery can charge while installed in a CR2700 reader using the inductive charging station (CRA-A271).
Caution: Model BRDY176699 has lithium-ion battery - must ship ground.
Specifications
- Description:Cable, 6' length, dark grey, for model CR2700 Inductive charging station (176514)
- Seq:01