0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
FORMAT® BLUE 1 * 100 ST
FORMAT® BLUE 1 * 100 ST
UNIG6002
:
UNIGLOVES AERTZE UND KLINIKBED
FORMAT® BLUE 1 * 100 ST
UNIG6002
:
UNIGLOVES AERTZE UND KLINIKBED
FORMAT® BLUE 1 * 100 ST
42,20 €
1 * 100 items
-
+