Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SENSOR PT100 -70 + - 600C 1 * 1 items
Catalog # CLEA626096
Supplier:
CleanAir by Baker
Expand image
SENSOR PT100 -70 + - 600C 1 * 1 items
Catalog # CLEA626096
Supplier:
CleanAir by Baker