Order Entry
Export
Orders LinkContactUsLinkComponent
 

Label Printer Kits, S3700

S3700 Multicolour and cut sign and label printer with AZERTY keyboard with BWS SFIDS, 101,6 mm (max), 482,60×361,95×279,40 mm
Catalog # BRDY321267
Supplier:   Brady
Label Printer Kits, S3700
Catalog # BRDY321267
Supplier:  Brady
Label Printer Kits, S3700
Label Printer Kits, S3700

S3700 Multicolour and cut sign and label printer with AZERTY keyboard with BWS SFIDS, 101,6 mm (max), 482,60×361,95×279,40 mm

Price
3 650,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Multicolour, cut sign and label printers, S3700 with QWERTZ keyboard, Wifi, bluetooth enabled and BWS SFIDS included. Colour your way to an improved workplace with the S3700 Sign and Label Printer. With multicolour print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.

  • Multicolour print capabilities make it ideal for creating impactful safety signs, lockout tags, warning labels and more in a single pass
  • Built in XY-cutter allows for the creation of die-cut shapes and text that gives the flexibility to choose where to place new messaging
  • The built-in keyboard and touchscreen interface make label creation a breeze
  • Label creation is even faster with the quick setup and print speeds that come with print smart technology from Brady
  • Prints on a variety of material options so you can create signs and labels for a variety of applications
  • Includes Brady workstation safety and facility ID software suite to easily create custom signs, labels and pipemarkers

Applications: Asset tracking, barcode labelling, cold temperature labelling, component and equipment labelling, electrical labelling, exit and egress marking, facility identification, floor marking, food and beverage process labelling, general identification, hazard communication identification, healthcare and hospital labelling, inventory and inspection labelling, lean and 5S labelling, pipe and valve marking, safety identification, security identification, temperature indicating labelling, temporary labelling, warehouse marking.

Media wiper ensures clean finished labels and catches dust so your printer runs stronger. Media guides and integrated sensors automatically align when guides squeeze together - no trial-and-error adjustments. Precise XY plotter cutter provides smoother edges, more precise connection points, straighter lines and more shapes for every label.

Optimised multi-color printing: Multi-patch ribbons require less frequent changes, enabling faster printing, less material waste and optimal efficiency
Ribbon control with fine tuning: Ribbon cartridges last up to 60% longer thanks to smarter printing technology

Smart media reels: Brady technology automatically adjusts speed, size, heat, layout and ribbon/material combination
Easy label and ribbon change in 20 seconds: Easy to deploy - no threading, no sensor adjustment, no calibration and no wasted labels
Various label widths: Prints labels from 12,70 to 101,60 mm wide on a wide variety of highly durable materials

Connection options: Bluetooth® 5 Low Energy (class II), Ethernet, Standalone Use, USB, Wi-Fi
Software compatibility: Brady Workstation, Express Labels Mobile App, Windows based driver for 3rd party software use
Barcode type: Code 128 Auto, Code 39, EAN-13, Interleaved 2 of 5, More through Brady WorkStation, UPC-A
Integrated symbols: 664

Specifications
  • Print width:
    101,6 mm (max)
  • Description:
    S3700 Multicolour and cut sign and label printer with AZERTY keyboard with BWS SFIDS
  • Seq:
    02
  • W×D×H:
    482,60×361,95×279,40 mm
  • Cat. No.:
    BRDY321267
  • Print speed:
    127 mm/s
  • Print method:
    Thermal transfer
  • Print resolution:
    300 dpi
  • Power supply:
    AC adaptor
  • Electrical:
    110/240 V
  • Connector type:
    Bluetooth® 5 Low Energy (class II), Ethernet, Standalone Use, USB, Wi-Fi
  • Weight:
    16,74 kg
  • Display:
    Color LCD touchscreen
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Workstation lockout writer software app
    Workstation lockout writer software app
    BRDY145445
    BBP®31, BBP®33, BBP®85, i3300
    Log in for availability
    1 * 1 items - 833,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Brady workstation CLP/GHS app via download
    Brady workstation CLP/GHS app via download
    BRDY195526
    Log in for availability
    1 * 1 items - 369,99 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Brady workstation automation software suite
    Brady workstation automation software suite
    BRDY149187
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    Log in for availability
    1 * 1 items - 833,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Brady workstation scan and print software suite
    Brady workstation scan and print software suite
    BRDY149445
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    Log in for availability
    1 * 1 items - 833,00 €