Orders LinkContactUsLinkComponent
Tanks for water baths
Tanks for water baths
Plexiglas® tank, 345×140×155 mm
Catalog # HECH43194001
Tanks for water baths
Catalog # HECH43194001
Tanks for water baths
Tanks for water baths
Plexiglas® tank, 345×140×155 mm
Price
144,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Tubs made of Plexiglas®, available with or without sloping base. Sloping base facilitates circulation of the liquid.
For use with water bath thermostats.
Specifications
  • Description:
    Plexiglas® tank, 345×140×155 mm