ContactUsLinkComponent
Furnace MuFFle with scale anD soFtware lt9/11/sw/c550 tMax1100 with liFt Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABELW095H1ANOTZ
Supplier:  NABERTHERM
Furnace MuFFle with scale anD soFtware lt9/11/sw/c550 tMax1100 with liFt Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABELW095H1ANOTZ
Supplier:  NABERTHERM