ContactUsLinkComponent
Furnace MuFFle lt9/11/skM/b510 tMax1100 flap door and over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABELM095K1ANLTZ
Supplier:  NABERTHERM
Furnace MuFFle lt9/11/skM/b510 tMax1100 flap door and over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABELM095K1ANLTZ
Supplier:  NABERTHERM