ContactUsLinkComponent
Furnace MuFFle l5/13/b510 tMax1300 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABEL-055K3ANLTZ
Supplier:  NABERTHERM
Furnace MuFFle l5/13/b510 tMax1300 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABEL-055K3ANLTZ
Supplier:  NABERTHERM