ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 321 Midcap Filters configuration: 1P Back panel: transparent Front and Sid e panel: sequential opening+ standard Power supply: CH Version: 1 1 * 1 items
Catalog # ERLAAA602E00021A41
Supplier:  ERLAB CAPTAIRE
Model technology: Filtration ductless fume hood Captair 321 Midcap Filters configuration: 1P Back panel: transparent Front and Sid e panel: sequential opening+ standard Power supply: CH Version: 1 1 * 1 items
Catalog # ERLAAA602E00021A41
Supplier:  ERLAB CAPTAIRE