ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 321 Midcap Filters configuration: 1P Back panel: standard Front and Side p anel: sequential opening+ standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA602E00011A31
Supplier:  ERLAB CAPTAIRE
Model technology: Filtration ductless fume hood Captair 321 Midcap Filters configuration: 1P Back panel: standard Front and Side p anel: sequential opening+ standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA602E00011A31
Supplier:  ERLAB CAPTAIRE