Model technology: Filtration ductless fume hood Captair 321 Midcap Filters configuration: 1P Back panel: standard Front and Side p anel: sequential opening+ standard Power supply: EU Version: 1 1 * 1 items
Model technology: Filtration ductless fume hood Captair 321 Midcap Filters configuration: 1P Back panel: standard Front and Side p anel: sequential opening+ standard Power supply: EU Version: 1 1 * 1 items