Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Melting Point Caffeine 234 - 236,5 C 1 * 1 g
Catalog # KRUEKSPS1014
Supplier:
A.KRÜSS Optronic
Expand image
Melting Point Caffeine 234 - 236,5 C 1 * 1 g
Catalog # KRUEKSPS1014
Supplier:
A.KRÜSS Optronic