Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CE14
Supplier:
Sartorius Balances
Expand image
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CE14
Supplier:
Sartorius Balances