Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA36S3S00234M
Supplier:
Sartorius Balances
Expand image
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA36S3S00234M
Supplier:
Sartorius Balances