Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Canister 25 liter, S60/61, PE-HD, UN-Y approval, dimensions (W x H x D): 250 x 475 x 295 mm 1 * 1 items
Catalog # SCTV107755
Supplier:
S.C.A.T.
Expand image
Canister 25 liter, S60/61, PE-HD, UN-Y approval, dimensions (W x H x D): 250 x 475 x 295 mm 1 * 1 items
Catalog # SCTV107755
Supplier:
S.C.A.T.