Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Human Niemann Pick C1 ELISA Kit 1 * 96 Tests
Human Niemann Pick C1 ELISA Kit 1 * 96 Tests
Human Niemann Pick C1 ELISA Kit
Catalog # ABCAAB313912-96TST
Supplier:
Abcam
CAS Number:
Expand image
Human Niemann Pick C1 ELISA Kit
Catalog # ABCAAB313912-96TST
Supplier:
Abcam
CAS Number:
About this item
Product Details
Product Details