0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant mouse IL-12 protein (Active) 1 * 250 µG
ABCAAB259419-250UG
:
Abcam
Recombinant mouse IL-12 protein (Active) 1 * 250 µG
ABCAAB259419-250UG
:
Abcam