Mouse Interferon alpha 1 ELISA Kit
Mouse Interferon alpha 1 ELISA kit, 384 tests
Catalog # ABCAAB252352-1X384
Supplier: Abcam
Jump to:
Product Details & Documents
Mouse Interferon alpha 1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Interferon alpha 1 with a sensitivity of 7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations.
Specifications
Specifications
- Description:Mouse Interferon alpha 1 ELISA kit
- Tests per kit:384 tests
- Primary antibody reactivity:Mouse
- Sample type:Cell culture media, Heparin Plasma, Citrate plasma, Serum
- Detection range:15,6 - 1000 pg/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB252352-1X384
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:7 pg/ml
- Format:Pre-coated
- Target protein:Interferon alpha 1
- Regulatory status:RUO



