Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HPLC 200X4.6MM PERFECTSIL 100 ODS-3 3µM 1 * 1 ST
Catalog # MSPPMZ0708-200046
Supplier:
MARKET SOURCE PART PROCESS
Expand image
HPLC 200X4.6MM PERFECTSIL 100 ODS-3 3µM 1 * 1 ST
Catalog # MSPPMZ0708-200046
Supplier:
MARKET SOURCE PART PROCESS