Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
600 L
Supplier: Avantor
Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
Catalog # 2622.9600
Supplier: Avantor
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- UN:1823
- ADR:8,II
Documents
Specifications
Specification Test Results
- Assay (NaOH):49 - 51 %
- Appearance (clear, colorless liquid):passes test
- Potassium (K):max. 0.01 %
- Identification (Na):passes test
- Sodium Carbonate (Na2CO3):max. 1.0 %
- Trace Impurities (in ppm) Chlorides (as Cl):max. 5
- Trace Impurities (in ppm) Iron (Fe):max. 2
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 1
Specifications
- Pk:600 L



