Order Entry
Export
Orders LinkContactUsLinkComponent
 

Graphite, powder, GPR RECTAPUR®

PP bucket, 25 kg
Catalog # 24488.460
Supplier:   VWR Chemicals
Graphite, powder, GPR RECTAPUR®
Catalog # 24488.460
Supplier:  VWR Chemicals
CAS Number:  7782-42-5
Graphite, powder, GPR RECTAPUR®
Graphite, powder, GPR RECTAPUR®

PP bucket, 25 kg

CAS Number:  7782-42-5
Price
512,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2.09…2.23 g/cm³ (20 °C)
  • EINECS:231-955-3
Specifications

Specification Test Results

  • Loss on drying (120°C):
    ≤ 2.5 %
  • Particle size (< 75 μm):
    ≥ 95 %
  • Substances soluble in ethanol:
    ≤ 0.2 %

Specifications

  • Pack type:
    PP bucket
  • Pk:
    25 kg

Specifications