Graphite, powder, GPR RECTAPUR®
PE bag in carton, 1 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- CAS Number:7782-42-5
- Formula:C
- MDL Number:MFCD00144065
- Molecular Weight:12.01 g/mol
- Merck Index:12,04560
- Melting Pt:3827 °C
- Density:2.09…2.23 g/cm³ (20 °C)
- EINECS:231-955-3
Documents
Certificates
Search by Lot Numberspec_VWRC24488.290 spec_VWRC24488.360 spec_VWRC24488.460
Product specification
Specifications
Specification Test Results
- Loss on drying (120°C):≤ 2.5 %
- Particle size (< 75 μm):≥ 95 %
- Substances soluble in ethanol:≤ 0.2 %
Specifications
- Pack type:PE bag in carton
- Pk:1 kg



