Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
VIAL 1.7ML CLEAR SNAP 11MM WIDE OPENING INNER V-BOTTOM HIGH RECOVERY 100/PACK 1 * 100 items
HYPE6PRV11-1V
:
Thermo Fisher Scientific
VIAL 1.7ML CLEAR SNAP 11MM WIDE OPENING INNER V-BOTTOM HIGH RECOVERY 100/PACK 1 * 100 items
HYPE6PRV11-1V
:
Thermo Fisher Scientific