Oxygen Sensor, Memosens COS81E
Product Details & Documents
Memosens 2.0 hygienic sensor for the life sciences and food industries.
- Memosens COS81E is designed according to EHEDG and ASME BPE. It avoids cross-contamination and is fully compliant to USP class VI and FDA as well as all GMP and GLP requirements
- High process up-time: Precalibrate the sensor in your lab and then swap it into your process with plug and play. It does not need polarization time and is immediately ready to measure
- Reliable measurement: A built-in reference LED compensates the ageing of the measuring LED. This ensures precise measured values even after frequent CIP/SIP and autoclaving
- Reduce your maintenance effort: Memosens COS81E does not have a difficult-to-handle electrolyte or sensitive membrane. Just exchange the sensor cap, perform a calibration and you are done
- The sensor can be used in process applications as well as benchtop fermenters. Providing you with 100% measuring consistency from the first lab trials to the final scaled-up process and your process lab
- IIoT ready: Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
Memosens COS81E is the ideal optical oxygen sensor for hygienic applications. Its well-tuned measuring system prevents ageing of the optical spot and offers precise, long-term stable measurements for high product quality. With Memosens 2.0 digital technology, COS81E offers extended data storage providing the perfect basis for predictive maintenance and efficient operation.
Memosens COS81E measures oxygen in applications with high demands to sterility and hygiene such as fermentation processes, from lab to process.
Measurement range: 0 to 30 mg/L
Certifications: EN 61326-1 EN 61326-2-3; EN 50581; EN IEC 60079-0; UKCA; Nonhazardous area approval.
Accessories information: Maintenance service kit COV81 contains optical cap, O-rings and further material for maintenance of Memosens COS81E oxygen sensor.
Documents
Specifications
- Connection:Memosens
- Description:Memosens COS81E oxygen sensor, 320 mm sensor length, stainless steel C-shape cap, EPDM O-ring
- Seq:04
- Cat. No.:664-0246
- Sensor type:Optical dissolved oxygen
Accessories
Product Information
- Catalog #AvailabilityPrice
- COV81 Maintenance kit for COS81E, EPDM O-ringCOV81 Maintenance kit for COS81E, EPDM O-ring662-2469Log in for availability1 * 1 items - 671,00 €
Product Information
- Catalog #AvailabilityPrice
- COY8 Zero gel for oxygen sensors (cert. HE_01083_01.18)COY8 Zero gel for oxygen sensors (cert. HE_01083_01.18)662-2466Log in for availability1 * 25 mL - 51,60 €



