Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®

Plastic container, 10 kg
Catalog # 302426C
Supplier:   VWR Chemicals
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
Catalog # 302426C
Supplier:  VWR Chemicals
CAS Number:  50-70-4
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®

Plastic container, 10 kg

CAS Number:  50-70-4
Price
1 620,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:94…98 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    97.0- 100.5 %
  • Appearance of solution (10 %; water):
    Passes test
  • Identification:
    Passes test
  • Spec. opt. rot.(10 %; borate; anhydrous):
    4.0 - 7.0 °
  • Reducing sugars (calculated as C6H12O6):
    Max. 0.2 %
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    Max. 1.5 %
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Ni (Nickel):
    Max. 1 ppm
  • Pb (Lead):
    Max. 0.5 ppm

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 kg

Specifications