0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
TARGET 3 1 * 1 items
TARGET 3 1 * 1 items
HITA893-4834
:
VWR Collection
TARGET 3 1 * 1 items
HITA893-4834
:
VWR Collection
TARGET 3 1 * 1 items
1 030,00 €
1 * 1 items
-
+