0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant Human Collagen VI alpha 2 GS 1 * 25 µG
NOVUH001292-P01-25
:
Novus Biologicals
Recombinant Human Collagen VI alpha 2 GS 1 * 25 µG
NOVUH001292-P01-25
:
Novus Biologicals