Order Entry
Export
Orders LinkContactUsLinkComponent
J.T.Baker® Endonuclease

J.T.Baker® Endonuclease

Plastic Bottle, 1.250 KU
Catalog # BAKR7108-40
Supplier:   Avantor
J.T.Baker® Endonuclease
Catalog # BAKR7108-40
Supplier:  Avantor
CAS Number:  
J.T.Baker®  Endonuclease
J.T.Baker® Endonuclease

Plastic Bottle, 1.250 KU

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1.250 KU
Product Details & Documents

J.T.Baker® Endonuclease Ultrapure Bioreagent is designed for the degradation of both single stranded and double stranded DNA and RNA. J.T.Baker® Endonuclease Ultrapure Bioreagent is used to ensure host cell DNA impurities are removed; driving process efficiency by lowering viscosity and preventing aggregation. J.T.Baker® Endonuclease is an enzyme based upon the native endonuclease of Serratia marcescens, enabling rapid clearance of residual DNA and RNA during the production and purification of both recombinant proteins and viral vectors. Non-animal origin. The purity of materials in non-negotiable. J.T.Baker® Endonuclease Ultrapure Bioreagent acts to degrade and eliminate extraneous genetic material, ensuring the pristine quality of your final product.

  • High purity, bioburden and endotoxin tested
  • Rapid clearance of residual DNA and RNA
  • Easily cleared in downstream processing, detectable by routine analytical methods
  • 0.2 µm filtered solution
  • Various size offerings
  • Proven performance with J.T.Baker® Workflow solutions

Appearance: Passes Test
Identification: Passes Test
Purity by SDS-PAGE: ≥ 90%
Activity: ≥ 250 U/µL
Specific Activity: ≥ 1.1 x 10^6 U/mg
Total Aerobic Microbial Count: ≤ 10 cfu/ 100,000U
Total Yeast and Mold Count: ≤ 10 cfu/ 100,000U
Endotoxin: For Information Only

Specifications

Specifications

  • Pack type:
    Plastic Bottle
  • Pk:
    1.250 KU
  • Cat. No.:
    BAKR7108-40
  • Storage conditions:
    -20 °C
  • Grade:
    Ultrapure Bioreagent
  • Enzyme name:
    Endonuclease
  • CAS No.:
    9025-65-4

Specifications