0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant mouse IL-23 protein (Active) 1 * 250 µG
ABCAAB259423-250
:
Abcam
Recombinant mouse IL-23 protein (Active) 1 * 250 µG
ABCAAB259423-250
:
Abcam