Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Single-Channel Pipettes, Mechanical, Variable Volume

Standard Line, 10 - 100 µl, ±1,6 - ±0,8 %, ≤0,8 - ≤0,2 %, Yellow
  613-7105
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
  613-7105
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume

Standard Line, 10 - 100 µl, ±1,6 - ±0,8 %, ≤0,8 - ≤0,2 %, Yellow

 
235,00 €
   
1 * 1 items
 

These lightweight Standard Line pipettes provide accuracy and precision. Their ergonomic features, such as the contoured shape of the handle and the soft spring system, deliver smooth and effortless pipetting, even during extensive periods of use.

  • Easily adjustable ejector height system to accommodate virtually all brands of tips
  • UV resistant and fully autoclavable
  • Easy and quick recalibration system
  • Colour coded shaft for easy volume identification
  • 5000 and 10000 μl version with shaft protected by filter
 
  • Colour code:
    Yellow
  • Volume:
    10 - 100 µl
  • Description:
    Standard Line
  • Seq:
    08
  • Accuracy:
    ±1,6 - ±0,8 %
  • Imprecision:
    ≤0,8 - ≤0,2 %
Accessories

 

  •  
    Description
     
     
  • VWRISP19476
    Filter for standard and EHP 5 and 10 ml pipettes
       
    1 * 10 items - 51,00 €

 

  •  
    Description
     
     
  • 613-7230
    Shelf clip for 1 single or multichannel pipette, Standard Line, EHP and UHP
       
    1 * 1 items - 15,20 €