VWR® PETG/PC Sample Bottles
20 ml, Sterile, 29,8 mm, 20-415
Catalog # VWRI210-000051
Supplier: VWR Collection
Jump to:
Product Details & Documents
VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.
- Made from durable, break-resistant 100% virgin PETG material
- Suitable for frozen storage down to −70°C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
Bottles are arranged in shrink-wrapped trays and polybag packaging to prevent contamination. Bottles 30 ml and above feature volume graduations. Non sterile bottles are autoclavable.
Certifications: US FDA 21CRF, USP Class VI compliant resin.
Accessories information: Spare closures are available for specific application requirements.
Packaging: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.
Documents
Certificates
Search by Lot NumberCoC VWR PC Bottles Extractables and Leachable CoC VWR® Media Bottles, Square, PETG coc_VWRI210-000043_238V-52-16J-210723 coc_VWRI210-000044_244V-52-16J-250723 coc_VWRI210-000045_249V-52-16J-270723
More
Specifications
- Cap Size:20-415
- Ø:29,8 mm
- Type:Sterile
- Capacity:20 ml
- Seq:09



