Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® PETG/PC Sample Bottles

VWR® PETG/PC Sample Bottles

5 ml, Sterile, 22,5 mm, 20-415
Catalog # VWRI210-000049
Supplier:   VWR Collection
VWR® PETG/PC Sample Bottles
Catalog # VWRI210-000049
Supplier:  VWR Collection
VWR® PETG/PC Sample Bottles
VWR® PETG/PC Sample Bottles

5 ml, Sterile, 22,5 mm, 20-415

Price
329,00 €
Log in for availability
1 * 100 items
Product Details & Documents

VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.

  • Made from durable, break-resistant 100% virgin PETG material
  • Suitable for frozen storage down to −70°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability

Bottles are arranged in shrink-wrapped trays and polybag packaging to prevent contamination. Bottles 30 ml and above feature volume graduations. Non sterile bottles are autoclavable.

Certifications: US FDA 21CRF, USP Class VI compliant resin.

Accessories information: Spare closures are available for specific application requirements.

Packaging: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.

Specifications
  • Cap Size:
    20-415
  • Ø:
    22,5 mm
  • Type:
    Sterile
  • Capacity:
    5 ml
  • Seq:
    07