Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethanol ≥96% (v/v), TechniSolv®, pure

950 L
  83804.930
 
 
Ethanol ≥96% (v/v), TechniSolv®, pure
  83804.930
 :  64-17-5
Ethanol ≥96% (v/v), TechniSolv®, pure
Ethanol ≥96% (v/v), TechniSolv®, pure

950 L

 :  64-17-5
 
 
   
1 * 950 L
 

 : WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

 :  Ethyl alcohol
  •  :64-17-5
  •  :C₂H₅OH
  •  :MFCD00003568
  •  :46.07 g/mol
  •  :78.3 °C (1013 hPa)
  •  :–117 °C
  •  :0.7895 g/cm³ (20 °C)
  •  :12 °C (closed cup)
  •  :Ambient
  •  :200-578-6
  •  :1170
  •  :3,II
  •  :01-2119457610-43
 

 

  • Assay:
    Min. 95 %
  • Assay (V/V):
    Min. 95 %
  • Density (20/20):
    0.805 - 0.811

 

  • Pk:
    950 L