ContactUsLinkComponent
10 ml - 10R Injection vial made of Type 1 Clear glass, with Crimp neck 20 mm neck finish. Pharmaceutical grade tubular glass. Dimens ions (OD x H) 24 mm x 45 mm. 1 * 174 items
Catalog # ALUG1008728
Supplier:  APG Europe
10 ml - 10R Injection vial made of Type 1 Clear glass, with Crimp neck 20 mm neck finish. Pharmaceutical grade tubular glass. Dimens ions (OD x H) 24 mm x 45 mm. 1 * 174 items
Catalog # ALUG1008728
Supplier:  APG Europe